CAS RN 1632006-28-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Esaxerenone, MedChemExpress
MedChemExpress Esaxerenone (CS-3150) is a highly potent and selective non-steroidal mineralocorticoid receptor antagonist.