CAS RN 163047-21-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Anemarrhenasaponin I, MedChemExpress
MedChemExpress Anemarrhenasaponin I, a traditional Chinese medicine, shows remarkable inhibiting effect on platelet aggregation.