CAS RN 1629063-81-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
GNE-131, MedChemExpress
MedChemExpress GNE-131 is a potent and selective inhibitor of human sodium channel NaV1.7, with an IC50 of 3 nM.