CAS RN 1628830-21-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
EZM 2302, MedChemExpress
MedChemExpress EZM 2302 is an inhibitor of coactivator-associated arginine methyltransferase 1 (CARM1) with an IC50 of 6 nM.