CAS RN 1628805-46-8
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
IDH-305, MedChemExpress
MedChemExpress IDH-305 is an orally available, mutant-selective and brain-penetrant IDH1 inhibitor that targets IDH1 (R132) mutation. IDH-305 exhibits greater than 200 fold selectivity for mutant IDH1 isoforms vs. WT (IC50= 27 nM (IDH1R132H), 28 nM (IDH1R132C), 6.14 μM (IDH1WT)).