CAS RN 1628428-01-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
PKR-IN-2, MedChemExpress
MedChemExpress PKR-IN-2 is a pyruvate kinase isoform PKR activator extracted from patent WO2014139144A1, compound 160. PKR-IN-2 can be used for the research of PKR function related diseases, including cancer, diabetes, obesity, autoimmune disorders, and benign prostatic hyperplasia.