CAS RN 162558-52-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
Tocris Bioscience™ MQAE
Fluorescent Cl- indicator; membrane permeable
2
MedChemExpress MQAE
MedChemExpress MQAE is a fluorescent indicator that is quenched via collision with chloride, and is more sensitive and selective than 36Cl and microelectrode-based methods for chloride measurement in cells.