CAS RN 161832-65-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Talampanel, MedChemExpress
MedChemExpress Talampanel (LY300164) is an orally and selective α-amino-3-hydroxy-5-methyl-4-isoxazolepropionate (AMPA) receptor antagonis with anti-seizure activity. Talampanel (IVAX) has neuroprotective effects in rodent stroke models. Talampanel attenuates caspase-3 dependent apoptosis in mouse brain.