CAS RN 1616392-22-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
GSK3326595, MedChemExpress
MedChemExpress GSK3326595 (EPZ015938) is a potent, selective, reversible inhibitor of protein arginine methyltransferase 5 (PRMT5) with an IC50 of 6.2 nM.