CAS RN 161605-73-8
IUPAC Nombre:
{[7-(morpholin-4-yl)-2,3-dioxo-6-(trifluoromethyl)-1,2,3,4-tetrahydroquinoxalin-1-yl]methyl}phosphonic acid
Sinónimos:
fanapanel
Peso molecular (g/mol):
409.26
Fórmula molecular:
C14H15F3N3O6P
InChi Key:
WZMQMKNCWDCCMT-UHFFFAOYSA-N
SMILES:
OP(O)(=O)CN1C(=O)C(=O)NC2=CC(=C(C=C12)N1CCOCC1)C(F)(F)F
1
–
2
de
2
resultados
1
Tocris Bioscience™ ZK 200775
CAS: 161605-73-8 Fórmula molecular: C14H15F3N3O6P
2
Fanapanel, MedChemExpress
MedChemExpress Fanapanel (ZK200775) is a highly selective AMPA/kainate antagonist with little activity against NMDA; have Ki values of 3.2 nM, 100 nM, and 8.5 μM against quisqualate, kainate, and NMDA, respectively.