CAS RN 160800-57-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Auristatin E, MedChemExpress
MedChemExpress Auristatin E is a cytotoxic tubulin modifier with potent and selective antitumor activity; MMAE analog and cytotoxin in Antibody-drug conjugates. Auristatin E inhibits cell division by blocking the polymerisation of tubulin.