CAS RN 159646-83-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
3
de
3
resultados
1
MedChemExpress QL9
MedChemExpress QL9 (QLSPFPFDL) is a high-affinity alloantigen for the 2C T cell receptor (TCR).
2
MedChemExpress QL9
MedChemExpress QL9 (QLSPFPFDL) is a high-affinity alloantigen for the 2C T cell receptor (TCR).
3
MedChemExpress QL9
MedChemExpress QL9 (QLSPFPFDL) is a high-affinity alloantigen for the 2C T cell receptor (TCR).