CAS RN 15866-90-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Incyclinide, MedChemExpress
MedChemExpress Incyclinide (CMT-3, COL-3) is a matrix metalloproteinase (MMP) inhibitor, thereby inducing extracellular matrix degradation, and inhibiting angiogenesis, tumor growth and invasion, and metastasis.