CAS RN 1579991-63-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Chitinase-IN-2, MedChemExpress
MedChemExpress Chitinase-IN-2 is a insect chitinase and N- acetyl hexosaminidase inhibitor and pesticide; 50 μM/20 μM compound concentration's inhibitory percentage are 98%/92% for chitinase/N- acetyl-hexosaminidase respectively.