CAS RN 156-39-8
IUPAC Nombre:
3-(4-hydroxyphenyl)-2-oxopropanoic acid
Sinónimos:
4-hydroxyphenylpyruvic acid (p-hydroxyphenyl)pyruvic acid (4-hydroxyphenyl)pyruvic acid
Peso molecular (g/mol):
180.16
Fórmula molecular:
C9H8O4
InChi Key:
KKADPXVIOXHVKN-UHFFFAOYSA-N
SMILES:
OC(=O)C(=O)CC1=CC=C(O)C=C1