CAS RN 1558021-37-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
TP0463518, MedChemExpress
MedChemExpress TP0463518 is a potent hypoxia-inducible factor prolyl hydroxylases (PHDs) inhibitor with a Ki value of 5.3 nM for human PHD2. TP0463518 also inhibits human PHD1/PHD3 with IC50s of 18 and 63 nM as well as monkey PHD2 with an IC50 value of 22 nM.