CAS RN 154992-24-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
RU 58841, MedChemExpress
MedChemExpress RU 58841 (PSK-3841) is a specific androgen receptor antagonist or anti-androgen. RU 58841 (PSK-3841) has a dramatic effect on hair regrowth.