CAS RN 153436-54-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
PD153035, MedChemExpress
MedChemExpress PD153035 (SU-5271; AG1517; ZM 252868) is a potent EGFR inhibitor with Ki and IC50 of 6 and 25 pM, respectively.