CAS RN 153436-53-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
AG-1478, MedChemExpress
MedChemExpress AG-1478 (Tyrphostin AG-1478) is a selective EGFR tyrosine kinase inhibitor with IC50 of 3 nM. AG-1478 has antiviral effects against HCV and encephalomyocarditis virus (EMCV).