CAS RN 152918-26-8
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
AB-MECA, MedChemExpress
MedChemExpress AB-MECA is a high affinity A3 adenosine receptor agonist, has high affinity for recombinant A1 and A3 receptors.