CAS RN 152751-57-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
Cloruro de Sevelamer (grado técnico), TRC
CAS: 152751-57-0 Fórmula molecular: (C3H7N)m• (C3H5ClO)n• (HCl)x Peso molecular (g/mol): 186.08 Sinónimo: 2-Propen-1-amine Hydrochloride polymer with 2-(Chloromethyl)oxirane,2-Propen-1-amine Hydrochloride polymer with (Chloromethyl)oxirane,(Chloromethyl)oxirane polymer with 2-Propen-1-amine Hydrochloride,Allylamine Hydrochloride-epichlorhydrin Copolymer,Allylamine Hydrochloride-epichlorohydrin Copolymer,GT 16-026A,Phosblock,RenaGel; SMILES: NCC=C.ClCC1OC1
2
Sevelamer hydrochloride, MedChemExpress
MedChemExpress Sevelamer hydrochloride is a phosphate binding drug used to treat hyperphosphatemia in patients with chronic kidney disease; consists of polyallylamine that is crosslinked with epichlorohydrin.