CAS RN 1515856-92-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
XEN445, MedChemExpress
MedChemExpress XEN445 is a potent and selective EL inhibitor(IC50=0.237 uM), that showed good ADME and PK properties, and demonstrated in vivo efficacy in raising plasma HDLc concentrations in mice.