CAS RN 15100-75-1
IUPAC Nombre:
hydrogen tert-butyl 2-amino-3-phenylpropanoate chloride
Sinónimos:
hydrogen tert-butyl 2-amino-3-phenylpropanoate chloride
Peso molecular (g/mol):
257.76
Fórmula molecular:
C13H20ClNO2
InChi Key:
FDMCEXDXULPJPG-UHFFFAOYNA-N
SMILES:
[H+].[Cl-].CC(C)(C)OC(=O)C(N)CC1=CC=CC=C1
1
–
1
de
1
resultados
1
Clorhidrato de tert-butil éster de L-fenilalanina, 98 %, Thermo Scientific Chemicals
CAS: 15100-75-1 Fórmula molecular: C13H20ClNO2 Peso molecular (g/mol): 257.758 Número MDL: MFCD00038894 Clave InChI: FDMCEXDXULPJPG-MERQFXBCSA-N Sinónimo: h-phe-otbu.hcl,h-phe-otbu hcl,tert-butyl 3-phenyl-l-alaninate hydrochloride,s-tert-butyl 2-amino-3-phenylpropanoate hydrochloride,l-phenylalanine tert-butyl ester hydrochloride,l-phenylalanine t-butyl ester hydrochloride,tert-butyl 2s-2-amino-3-phenylpropanoate hydrochloride,phe-otbu.hcl,tert-butyl l-phenylalaninate hydrochloride,phenylalanine, 1,1-dimethylethyl ester, hydrochloride PubChem CID: 11459563 Nombre IUPAC: tert-butil (2S)-2-amino-3-fenilpropanato;clorhidrato SMILES: CC(C)(C)OC(=O)C(CC1=CC=CC=C1)N.Cl