CAS RN 1505453-59-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
BET bromodomain inhibitor, MedChemExpress
MedChemExpress BET bromodomain inhibitor is a potent BET inhibitor extracted from patent WO/2015/153871A2, compound example 11.