CAS RN 1488354-15-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Fulacimstat, MedChemExpress
MedChemExpress Fulacimstat is an orally available chymase inhibitor, with IC50s of 4, 3 nM for human and hamster chymase enzyme, respectively.