CAS RN 148717-90-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Squalamine, MedChemExpress
MedChemExpress Squalamine(MSI-1256) is an aminosterol compound with potent broad spectrum antiviral activity.