CAS RN 14782-19-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
INO5042, MedChemExpress
MedChemExpress INO5042, a thiazole fused 1,4-naphthoquinone compound, and exhibits anti-inflammation activity.