CAS RN 147254-64-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Ranirestat, MedChemExpress
MedChemExpress Ranirestat (AS-3201) potent and orally active aldose reductase (AR) inhibitor with IC50s of 11 nM and 15 nM for rat lens AR and recombinant human AR, respectively, and a Ki of 0.38 nM for recombinant human AR. Ranirestat has the potential for diabetic sensorimotor polyneuropathy treatment. Ranirestat also has a neuroprotective effect on diabetic retinas.