CAS RN 1456858-58-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
HG-9-91-01, MedChemExpress
MedChemExpress HG-9-91-01 is a potent and highly selective salt-inducible kinase (SIK) inhibitor with IC50s of 0.92 nM, 6.6 nM and 9.6 nM for SIK1, SIK2 and SIK3 respectively.