CAS RN 145317-11-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Bisindolylmaleimide X hydrochloride, MedChemExpress
MedChemExpress Bisindolylmaleimide X hydrochloride (BIM-X hydrochloride) is a potent and selective protein kinase C (PKC) inhibitor. Bisindolylmaleimide X hydrochloride is a potent cyclin-dependent kinase 2 (CDK2) antagonist with an IC50 of 200 nM.