CAS RN 1449578-65-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
AZ82, MedChemExpress
MedChemExpress AZ82 is a selective kinesin-like protein KIFC1 (HSET/KIFC1) inhibitor, with a Ki of 43 nM and an IC50 of 300 nM for KIFC1.