CAS RN 1446817-84-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
ABX-1431, MedChemExpress
MedChemExpress ABX-1431 is a highly potent, selective, and orally available, CNS-penetrant monoacylglycerol lipase (MAGL) inhibitor with an IC50 of 14 nM.