CAS RN 1445251-22-8
IUPAC Nombre:
3-(4-oxo-1,4-dihydroquinazolin-2-yl)-N-(1-phenylethyl)propanamide
Sinónimos:
3-(4-oxo-1H-quinazolin-2-yl)-N-(1-phenylethyl)propanamide
Peso molecular (g/mol):
321.38
Fórmula molecular:
C19H19N3O2
InChi Key:
QIHBWVVVRYYYRO-UHFFFAOYNA-N
SMILES:
CC(NC(=O)CCC1=NC(=O)C2=CC=CC=C2N1)C1=CC=CC=C1
1
–
2
de
2
resultados
1
2
ME0328, MedChemExpress
MedChemExpress ME0328 is a potent and selective ARTD3/PARP3 inhibitor with an IC50 of 0.89±0.28 μM.