CAS RN 1443748-47-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
3
de
3
resultados
1
Tocris Bioscience™ VU 0469650 Hydrochloride
Potent and selective negative allosteric modulator of mGlu1
2
Tocris Bioscience™ VU 0469650 Hydrochloride
Potent and selective negative allosteric modulator of mGlu1
3
Tocris Bioscience™ VU 0469650 Hydrochloride
Potent and selective negative allosteric modulator of mGlu1