CAS RN 1438391-30-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
CC-401 hydrochloride, MedChemExpress
MedChemExpress CC-401 hydrochloride is a potent inhibitor of all three forms of JNK with Ki of 25 to 50 nM.