CAS RN 1431697-90-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
SB-408124 Hydrochloride, MedChemExpress
MedChemExpress SB-408124 Hydrochloride is a selective non-peptide orexin receptor 1 (OX1) receptor antagonist with Kis of 57 nM and 27 nM in whole cell and membrane, respectively. SB-408124 Hydrochloride exhibits 50-fold selectivity over OX2 receptor.