CAS RN 1429239-98-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
CB1-IN-1, MedChemExpress
MedChemExpress CB1-IN-1 (BPRCB1184) is a peripherally restricted CB1R antagonist, with Ki of 0.3 nM and 21 nM for CB1R (EC50 = 3 nM) and CB2R, respectively.