CAS RN 1425381-60-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Gusacitinib, MedChemExpress
MedChemExpress Gusacitinib (ASN-002) is an orally active and potent dual inhibitor of spleen tyrosine kinase (SYK) and janus kinase (JAK) with IC50 values of 5-46 nM. Gusacitinib has anti-cancer activity in both solid and hematological tumor types.