CAS RN 1423715-09-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
SP2509, MedChemExpress
MedChemExpress SP2509 is a potent and selective antagonist of lysine specific demethylase 1 (LSD1) with an IC50 of 13 nM.