CAS RN 1420290-99-8
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
HJC0152 hydrochloride, MedChemExpress
MedChemExpress HJC0152 hydrochloride is a signal transducers and activators of transcription 3 (STAT3) inhibitor.