CAS RN 1418013-75-8
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
Tocris Bioscience™ NMS 873
Potent and selective p97 ATPase (VCP) allosteric inhibitor
2
NMS-873, MedChemExpress
MedChemExpress NMS-873 is a potent, selective allosteric VCP/p97 inhibitor with an IC50 value of 30 nM.