CAS RN 1415800-43-9
IUPAC Nombre:
N-phenyl-2,5-bis[4-(pyrrolidin-1-yl)piperidine-1-carbonyl]aniline
Sinónimos:
N-phenyl-2,5-bis[4-(pyrrolidin-1-yl)piperidine-1-carbonyl]aniline
Peso molecular (g/mol):
529.73
Fórmula molecular:
C32H43N5O2
InChi Key:
PQOOIERVZAXHBP-UHFFFAOYSA-N
SMILES:
O=C(N1CCC(CC1)N1CCCC1)C1=CC(NC2=CC=CC=C2)=C(C=C1)C(=O)N1CCC(CC1)N1CCCC1
1
–
1
de
1
resultados
1
UNC1215, MedChemExpress
MedChemExpress UNC1215 is a potent and selective inhibitor for the methyllysine (Kme) reading domain function of L3MBTL3 with a Kd value of 120 nM and an IC50 of 40 nM. UNC1215 has the potential to treat malignant brain tumor.