CAS RN 1411977-95-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Acetylene-linker-Val-Cit-PABC-MMAE, MedChemExpress
MedChemExpress Acetylene-linker-Val-Cit-PABC-MMAE (LCB14-0602) consists the ADCs linker (Acetylene-linker-Val-Cit-PABC) and potent tubulin inhibitor (MMAE). Acetylene-linker-Val-Cit-PABC-MMAE (LCB14-0602) is a drug-linker conjugate for ADC.