CAS RN 141-22-0
IUPAC Nombre:
(9Z,12R)-12-hydroxyoctadec-9-enoic acid
Sinónimos:
ricinoleic acid l'acide ricinoleique acide ricinoleique kyselina ricinolova ricinolic acid 12-hydroxyoleic acid (r-(e))-isomer ricinoleic acid ricinoleic acid preparation ricinolsaeure ricinic acid
Peso molecular (g/mol):
298.47
Fórmula molecular:
C18H34O3
InChi Key:
WBHHMMIMDMUBKC-QJWNTBNXSA-N
SMILES:
CCCCCC[C@@H](O)C\C=C/CCCCCCCC(O)=O
1
–
1
de
1
resultados
1
Ácido ricinólico (~80%), TRC
CAS: 141-22-0 Fórmula molecular: C18 H34 O3 Peso molecular (g/mol): 298.46 Sinónimo: 9-Octadecenoic acid, 12-hydroxy-, (9Z,12R)-,9-Octadecenoic acid, 12-hydroxy-, [R-(Z)]-,Ricinoleic acid (8CI),(9Z,12R)-12-Hydroxy-9-octadecenoic acid,(9Z,12R)-12-Hydroxyoctadec-9-enoic acid,(R)-12-Hydroxyoleic acid,(R)-Ricinoleic acid,12-D-Hydroxy-9-cis-octadecenoic acid,12-Hydroxy-cis-9-octadecenoic acid,12-Hydroxyoctadeca-9-enoic acid,CS 80,Cenwax C,Edenor Ri 90,H 1657,NSC 281242,Nouracid CS 80,Ricinic acid,Ricinolic acid Nombre IUPAC: (Z,12R)-12-hidroxioctadeco-9-ácido enoico SMILES: CCCCCC[C@@H](O)C\C=C/CCCCCCCC(=O)O