CAS RN 140926-75-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
NSC5844, MedChemExpress
MedChemExpress NSC5844 (RE-640) is a 4-aminoquinoline derivative, with antitumor and antimalarial activity.