CAS RN 140898-91-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Hexaminolevulinate hydrochloride, MedChemExpress
MedChemExpress Hexaminolevulinate hydrochloride is a fluorescent agent, has approved for cystoscopic detection of papillary bladder cancer.