CAS RN 140686-92-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
N-563, MedChemExpress
MedChemExpress N-563 is an analogue of deoxyspergualin with an immunostimulating activity,it promotes resistance to Candida albicans infection in mice.