CAS RN 1403783-31-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Nexturastat A, MedChemExpress
MedChemExpress Nexturastat A is a potent HDAC6 inhibitor with an IC50 of 5 nM. Nexturastat A also inhibits HDAC10 and metallo-β-lactamase domain-containing protein 2 (MBLAC2).