CAS RN 1403764-72-6
IUPAC Nombre:
2-methoxy-N-(3-methyl-2-oxo-1,2,3,4-tetrahydroquinazolin-6-yl)benzene-1-sulfonamide
Sinónimos:
2-methoxy-N-(3-methyl-2-oxo-1,4-dihydroquinazolin-6-yl)benzenesulfonamide
Peso molecular (g/mol):
347.39
Fórmula molecular:
C16H17N3O4S
InChi Key:
TXZPMHLMPKIUGK-UHFFFAOYSA-N
SMILES:
COC1=CC=CC=C1S(=O)(=O)NC1=CC2=C(NC(=O)N(C)C2)C=C1
1
–
2
de
2
resultados
1
Tocris Bioscience™ PFI 1
CAS: 1403764-72-6 Fórmula molecular: C16H17N3O4S
2
PFI-1, MedChemExpress
MedChemExpress PFI-1 is a selective BET (bromodomain-containing protein) inhibitor for BRD4 with IC50 of 0.22 μM in a cell-free assay.