CAS RN 1402821-41-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
GV-58, MedChemExpress
MedChemExpress GV-58 is a potent, selective N- and P/Q-type Ca2+ channels agonist with EC50 of 7.21/8.81 uM for N-type/P-Q-type Ca2+ channel; 20-fold less potent CDK inhibitor activity.